Anti-ATG16L1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP1817CEA 360 EUR
ABGEAP1817C
Anti-ATG16L1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ATG16L1 (autophagy related 16-like 1 (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR