Anti-ATG7 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP1813AEA 360 EUR
ABGEAP1813A
Anti-ATG7 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ATG7 (autophagy related 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR