Anti-ATG5 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP1812BEA 348 EUR
ABGEAP1812B
Anti-ATG5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ATG5 (autophagy related 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR