Anti-ATG4C Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP1810AEA 348 EUR
ABGEAP1810A
Anti-ATG4C Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ATG4C (autophagy related 4C, cysteine peptidase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR