Anti-LC3 (APG8B) Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP1802BEA 348 EUR
ABGEAP1802B
Anti-LC3 (APG8B) Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LC3 (APG8B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR