Anti-PTDSS1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP17127BEA 348 EUR
ABGEAP17127B
Anti-PTDSS1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PTDSS1 (phosphatidylserine synthase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR