Anti-FBXL13 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP17121BEA 348 EUR
ABGEAP17121B
Anti-FBXL13 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FBXL13 (F-box and leucine-rich repeat protein 13)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR