Anti-FAM65B Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP17120BEA 348 EUR
ABGEAP17120B
Anti-FAM65B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FAM65B (family with sequence similarity 65, member B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR