Anti-NCAPG2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP17069CEA 350 EUR
ABGEAP17069C
Anti-NCAPG2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NCAPG2 (non-SMC condensin II complex, subunit G2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR