Anti-SUPT7L Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP17064CEA 350 EUR
ABGEAP17064C
Anti-SUPT7L Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SUPT7L (suppressor of Ty 7 (S. cerevisiae)-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR