Anti-GNA13 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP17044CEA 350 EUR
ABGEAP17044C
Anti-GNA13 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GNA13 (guanine nucleotide binding protein (G protein), alpha 13)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR