Anti-SLPI Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP17015CEA 350 EUR
ABGEAP17015C
Anti-SLPI Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SLPI (secretory leukocyte peptidase inhibitor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR