Anti-HEYL Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP16997AEA 350 EUR
ABGEAP16997A
Anti-HEYL Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HEYL (hes-related family bHLH transcription factor with YRPW motif-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR