Anti-CAMP Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP16981BEA 348 EUR
ABGEAP16981B
Anti-CAMP Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CAMP (cathelicidin antimicrobial peptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR