Anti-YIPF3 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP16954BEA 350 EUR
ABGEAP16954B
Anti-YIPF3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: YIPF3 (Yip1 domain family, member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR