Anti-PTP4A1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP16923BEA 350 EUR
ABGEAP16923B
Anti-PTP4A1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PTP4A1 (protein tyrosine phosphatase type IVA, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR