Anti-RARB Rabbit Polyclonal Antibody

Lieferant: Abcepta
ABGEAP16899AEA 348 EUR
ABGEAP16899A
Anti-RARB Rabbit Polyclonal Antibody
Antikörper
LEAF (Low Endotoxin, Azide-Free)

Western Blot: 1:1000

Type: Primary
Antigen: RARB (retinoic acid receptor, beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR