Anti-MOB3B Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP16893AEA 350 EUR
ABGEAP16893A
Anti-MOB3B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MOB3B (MOB kinase activator 3B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR