Anti-WTAP Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP16885BEA 350 EUR
ABGEAP16885B
Anti-WTAP Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: WTAP (Wilms tumor 1 associated protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR