Anti-BOLL Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP16329AEA 350 EUR
ABGEAP16329A
Anti-BOLL Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: BOLL (boule-like RNA-binding protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR