Anti-ELOVL4 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP16326BEA 350 EUR
ABGEAP16326B
Anti-ELOVL4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ELOVL4 (ELOVL fatty acid elongase 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR