Anti-HSF5 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP16315BEA 350 EUR
ABGEAP16315B
Anti-HSF5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HSF5 (heat shock transcription factor family member 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR