Anti-CHCHD10 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP16303AEA 350 EUR
ABGEAP16303A
Anti-CHCHD10 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CHCHD10 (coiled-coil-helix-coiled-coil-helix domain containing 10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR