Anti-RBM45 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP16299BEA 348 EUR
ABGEAP16299B
Anti-RBM45 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RBM45 (RNA binding motif protein 45)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR