Anti-GBX1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP16293CEA 350 EUR
ABGEAP16293C
Anti-GBX1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GBX1 (gastrulation brain homeobox 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR