Anti-SLC39A14 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP16286AEA 360 EUR
ABGEAP16286A
Anti-SLC39A14 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SLC39A14 (solute carrier family 39 (zinc transporter), member 14)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR