Anti-SLAMF9 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP16284BEA 350 EUR
ABGEAP16284B
Anti-SLAMF9 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SLAMF9 (SLAM family member 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR