Anti-PAQR7 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP16283BEA 348 EUR
ABGEAP16283B
Anti-PAQR7 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PAQR7 (progestin and adipoQ receptor family member VII)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR