Anti-SIGLEC6 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP1624BEA 348 EUR
ABGEAP1624B
Anti-SIGLEC6 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SIGLEC6 (sialic acid binding Ig-like lectin 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR