Anti-OR4K13 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP16236AEA 350 EUR
ABGEAP16236A
Anti-OR4K13 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: OR4K13 (olfactory receptor, family 4, subfamily K, member 13)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR