Anti-XKR3 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP16213BEA 350 EUR
ABGEAP16213B
Anti-XKR3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: XKR3 (XK, Kell blood group complex subunit-related family, member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR