Anti-RHOG Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP16200BEA 348 EUR
ABGEAP16200B
Anti-RHOG Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RHOG (ras homolog family member G)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR