Anti-THAP11 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP16197AEA 350 EUR
ABGEAP16197A
Anti-THAP11 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: THAP11 (THAP domain containing 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR