Anti-SMARCD1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP16186BEA 360 EUR
ABGEAP16186B
Anti-SMARCD1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SMARCD1 (SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR