Anti-ZDHHC3 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP16170AEA 350 EUR
ABGEAP16170A
Anti-ZDHHC3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ZDHHC3 (zinc finger, DHHC-type containing 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR