Anti-NR2C1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP16146BEA 348 EUR
ABGEAP16146B
Anti-NR2C1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NR2C1 (nuclear receptor subfamily 2, group C, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR