Anti-SOX18 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP16137AEA 360 EUR
ABGEAP16137A
Anti-SOX18 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SOX18 (SRY (sex determining region Y)-box 18)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR