Anti-ITGB7 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP16093BEA 348 EUR
ABGEAP16093B
Anti-ITGB7 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ITGB7 (integrin, beta 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR