Anti-NLGN4Y Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP16091AEA 350 EUR
ABGEAP16091A
Anti-NLGN4Y Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NLGN4Y (neuroligin 4, Y-linked)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR