Anti-HOXD13 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP16079CEA 350 EUR
ABGEAP16079C
Anti-HOXD13 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HOXD13 (homeobox D13)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR