Anti-HOXB8 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP16065BEA 350 EUR
ABGEAP16065B
Anti-HOXB8 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HOXB8 (homeobox B8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR