Anti-C4BPB Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP16055CEA 350 EUR
ABGEAP16055C
Anti-C4BPB Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: C4BPB (complement component 4 binding protein, beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR