Anti-LTBR Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP16019AEA 348 EUR
ABGEAP16019A
Anti-LTBR Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LTBR (lymphotoxin beta receptor (TNFR superfamily, member 3))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR