Anti-P2RX2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP16011AEA 350 EUR
ABGEAP16011A
Anti-P2RX2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: P2RX2 (purinergic receptor P2X, ligand-gated ion channel, 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR