Anti-FAU Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP1600BEA 348 EUR
ABGEAP1600B
Anti-FAU Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FAU (Finkel-Biskis-Reilly murine sarcoma virus (FBR-MuSV) ubiquitously expressed)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR