Anti-TCEB2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP16007AEA 350 EUR
ABGEAP16007A
Anti-TCEB2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TCEB2 (transcription elongation factor B (SIII), polypeptide 2 (18kDa, elongin B))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR