Anti-GJD2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP1549BEA 350 EUR
ABGEAP1549B
Anti-GJD2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GJD2 (gap junction protein, delta 2, 36kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR