Anti-GJA3 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP1543AEA 348 EUR
ABGEAP1543A
Anti-GJA3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GJA3 (gap junction protein, alpha 3, 46kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR