Anti-STAT2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP1532AEA 348 EUR
ABGEAP1532A
Anti-STAT2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: STAT2 (signal transducer and activator of transcription 2, 113kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR