Anti-STAT5B Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP1530AEA 350 EUR
ABGEAP1530A
Anti-STAT5B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: STAT5B (signal transducer and activator of transcription 5B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR